Methyl-2,5,6-trichloro-4-pyrimidinecarboxylate 97%

Leverantör: Apollo Scientific
Varning

Synonyms: Methyl 2,5,6-trichloropyrimidine-4-carboxylate

OR33577-1G
APOSOR33577-1GEA 1450 SEK
APOSOR33577-1G
Methyl-2,5,6-trichloro-4-pyrimidinecarboxylate 97%
Methyl-2,5,6-trichloro-4-pyrimidinecarboxylate
Formel: C₆H₃Cl₃N₂O₂
MW: 241.46 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12025876
CAS nummer: 89284-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR