Methyl-1-(tert-butyl)-5-methyl-1H-pyrazole-4-carboxylate 95%
Leverantör: Apollo Scientific
OR33552-1G
OR33552-5G
APOSOR33552-1GEA
3525
SEK
APOSOR33552-1G
APOSOR33552-5G
Methyl-1-(tert-butyl)-5-methyl-1H-pyrazole-4-carboxylate 95%
Methyl-1-(tert-butyl)-5-methyl-1H-pyrazole-4-carboxylate
Formel:
C₁₀H₁₆N₂O₂ MW: 196,25 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD16140308 CAS nummer: 950858-97-6 |