3-(4-Morpholinophenyl)-3-oxopropanenitrile 95%
Leverantör: Apollo Scientific
OR33525-1G OR33525-5G
APOSOR33525-1GEA 3525 SEK
APOSOR33525-1G APOSOR33525-5G
3-(4-Morpholinophenyl)-3-oxopropanenitrile 95%
3-(4-Morpholinophenyl)-3-oxopropanenitrile
Formel: C₁₃H₁₄N₂O₂
MW: 230,27 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07787044
CAS nummer: 887591-40-4