3-Oxo-3-[4-(1H-pyrazol-1-yl)phenyl]propanenitrile 95%
Leverantör: Apollo Scientific
OR33476-1G OR33476-5G
APOSOR33476-1GEA 3950 SEK
APOSOR33476-1G APOSOR33476-5G
3-Oxo-3-[4-(1H-pyrazol-1-yl)phenyl]propanenitrile 95%
3-Oxo-3-[4-(1H-pyrazol-1-yl)phenyl]propanenitrile
Formel: C₁₂H₉N₃O
Storage Temperature: Omgivande
MDL Number: MFCD12025838
CAS nummer: 1176541-33-5