Thiosalicylic acid 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
2-Thiobenzoic acid, 2-Mercaptobenzoic acid
OR3344-1KG
OR3344-250G
APOSOR3344-1KGEA
14625
SEK
APOSOR3344-1KG
APOSOR3344-250G
Thiosalicylic acid 98%
Thiosalicylic acid
Formel:
C₇H₆O₂S MW: 154,19 g/mol Smältpunkt: 165…168 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00004836 CAS nummer: 147-93-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...