1-(3-Furyl)-1-ethanol 95%
Leverantör: Apollo Scientific
OR33378-1G
OR33378-5G
APOSOR33378-1GEA
3525
SEK
APOSOR33378-1G
APOSOR33378-5G
1-(3-Furyl)-1-ethanol 95%
1-(3-Furyl)-1-ethanol
Formel:
C₆H₈O₂ MW: 112,13 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD02082436 CAS nummer: 13129-26-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...