2'-Cyanoacetanilide 95%

Leverantör: Apollo Scientific

Synonyms: 2-Cyanoacetanilide, 2-Cyano-N-phenylacetamide

OR33320-1G OR33320-5G
APOSOR33320-1GEA 635 SEK
APOSOR33320-1G APOSOR33320-5G
2'-Cyanoacetanilide 95%
2'-Cyanoacetanilide
Formel: C₉H₈N₂O
MW: 160,1726 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00019840
CAS nummer: 621-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR