4-(1H-1,2,3-Benzotriazol-1-ylmethyl)phenylamine 95%
Leverantör: Apollo Scientific
Synonyms: 4-[(1H-1,2,3-Benzotriazol-1-yl)methyl]aniline
OR33288-1G OR33288-5G
APOSOR33288-1GEA 5225 SEK
APOSOR33288-1G APOSOR33288-5G
4-(1H-1,2,3-Benzotriazol-1-ylmethyl)phenylamine 95%
4-(1H-1,2,3-Benzotriazol-1-ylmethyl)phenylamine
Formel: C₁₃H₁₂N₄
MW: 224,27 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00960990
CAS nummer: 129075-89-4