2-(3,5-Dimethyl-1H-pyrazol-1-yl)-1-ethanol 95%
Leverantör: Apollo Scientific
OR33224-1G OR33224-25G OR33224-5G
APOSOR33224-1GEA 1550 SEK
APOSOR33224-1G APOSOR33224-25G APOSOR33224-5G
2-(3,5-Dimethyl-1H-pyrazol-1-yl)-1-ethanol 95%
2-(3,5-Dimethyl-1H-pyrazol-1-yl)-1-ethanol
Formel: C₇H₁₂N₂O
MW: 140,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00141692
CAS nummer: 20000-80-0