5-Amino-6-methylquinoline 95%

Leverantör: Apollo Scientific

Synonyms: 6-Methylquinolin-5-amine

OR32895-1G OR32895-5G
APOSOR32895-1GEA 2075 SEK
APOSOR32895-1G APOSOR32895-5G
5-Amino-6-methylquinoline 95%
5-Amino-6-methylquinoline
Formel: C₁₀H₁₀N₂
MW: 158,1998 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00094194
CAS nummer: 50358-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR