5-(Chloromethyl)-3-(4-methoxyphenyl)isoxazole 95%
Leverantör: Apollo Scientific
OR32870-1G
OR32870-5G
APOSOR32870-1GEA
5225
SEK
APOSOR32870-1G
APOSOR32870-5G
5-(Chloromethyl)-3-(4-methoxyphenyl)isoxazole 95%
5-(Chloromethyl)-3-(4-methoxyphenyl)isoxazole
Formel:
C₁₁H₁₀ClNO₂ MW: 223,66 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD01935948 CAS nummer: 325744-41-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...