6,7-Dihydro-4(5H)-benzofuranone 95%

Leverantör: Apollo Scientific

Synonyms: 6,7-Dihydrobenzofuran-4(5H)-one

OR32842-1G OR32842-5G
APOSOR32842-1GEA 685 SEK
APOSOR32842-1G APOSOR32842-5G
6,7-Dihydro-4(5H)-benzofuranone 95%
6,7-Dihydro-4(5H)-benzofuranone
Formel: C₈H₈O₂
MW: 136,15 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00215511
CAS nummer: 16806-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR