5-Bromo-3-methylbenzene-1,2-diamine 97%

Leverantör: Apollo Scientific

OR32811-1G OR32811-25G OR32811-5G
APOSOR32811-1GEA 685 SEK
APOSOR32811-1G APOSOR32811-25G APOSOR32811-5G
5-Bromo-3-methylbenzene-1,2-diamine 97%
5-Bromo-3-methylbenzene-1,2-diamine
Formel: C₇H₉BrN₂
MW: 201,0600 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06411036
CAS nummer: 76153-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR