6-(tert-Butyl)-2H-1,4-benzoxazin-3(4H)-one 95%
Leverantör: Apollo Scientific
OR32810-1G OR32810-5G
APOSOR32810-1GEA 3525 SEK
APOSOR32810-1G APOSOR32810-5G
6-(tert-Butyl)-2H-1,4-benzoxazin-3(4H)-one 95%
6-(tert-Butyl)-2H-1,4-benzoxazin-3(4H)-one
Formel: C₁₂H₁₅NO₂
MW: 205,26 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03830266
CAS nummer: 6238-96-6