1-(4-Chlorophenyl)-2-hydroxy-1-ethanone 95%
Leverantör: Apollo Scientific
Varning
OR32799-1G OR32799-25G OR32799-5G
APOSOR32799-1GEA 2075 SEK
APOSOR32799-1G APOSOR32799-25G APOSOR32799-5G
1-(4-Chlorophenyl)-2-hydroxy-1-ethanone 95%
1-(4-Chlorophenyl)-2-hydroxy-1-ethanone
Formel: C₈H₇ClO₂
MW: 170,6 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01570063
CAS nummer: 27993-56-2