8-Methoxy-2H-chromene-3-carbonitrile 97%
Leverantör: Apollo Scientific
OR32722-1G OR32722-25G OR32722-5G
APOSOR32722-1GEA 2125 SEK
APOSOR32722-1G APOSOR32722-25G APOSOR32722-5G
8-Methoxy-2H-chromene-3-carbonitrile 97%
8-Methoxy-2H-chromene-3-carbonitrile
Formel: C₁₁H₉NO₂
MW: 187.2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00202017
CAS nummer: 57543-69-8