1-Isopropenyl-2-benzimidazolidinone 97%
Leverantör: Apollo Scientific
Synonyms: 1-(Prop-1-en-2-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one, 1-(Prop-1-en-2-yl)-1H-benzo[d]imidazol-2(3H)-one
OR32517-5G OR32517-25G
APOSOR32517-5GEA 2850 SEK
APOSOR32517-5G APOSOR32517-25G
1-Isopropenyl-2-benzimidazolidinone 97%
1-Isopropenyl-2-benzimidazolidinone
Formel: C₁₀H₁₀N₂O
MW: 174,203 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00218253
CAS nummer: 52099-72-6