2-Acetylbenzothiazole 95%

Leverantör: Apollo Scientific

Synonyms: 1-(Benzo[d]thiazol-2-yl)ethanone, 1-(1,3-Benzothiazol-2-yl)ethanone

OR32487-1G OR32487-5G
APOSOR32487-1GEA 705 SEK
APOSOR32487-1G APOSOR32487-5G
2-Acetylbenzothiazole 95%
2-Acetylbenzothiazole
Formel: C₉H₇NOS
MW: 177.23 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00192994
CAS nummer: 1629-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR