4-tert-Butylkatekol

Leverantör: Apollo Scientific

Synonyms: 4-(tert-Butyl)benzene-1,2-diol, 4-tert-Butylcatechol

OR323288-1KG OR323288-250G OR323288-5KG
APOSOR323288-1KGEA 2075 SEK
APOSOR323288-1KG APOSOR323288-250G APOSOR323288-5KG
4-tert-Butylkatekol
4-tert-Butylkatekol
Formel: C₁₀H₁₄O₂
MW: 166,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00002201
CAS nummer: 98-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR