N-(Ethoxycarbonyl)phthalimide

Leverantör: Apollo Scientific

Synonyms: Ethyl 1,3-dioxoisoindoline 2-carboxylate, N-Carbethoxyphthalimide

OR323267-100G OR323267-25G
APOSOR323267-100GEA 1425 SEK
APOSOR323267-100G APOSOR323267-25G
N-(Ethoxycarbonyl)phthalimide
N-(Ethoxycarbonyl)phthalimide
Formel: C₁₁H₉NO₄
MW: 219,20 g/mol
Smältpunkt: 90…92 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00005893
CAS nummer: 22509-74-6
EINECS: 245-048-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR