(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulphonate salt 97%
Leverantör: Apollo Scientific
Fara
Synonyms: (S)-(-)-Tetrahydro-3-furylamine p-Toluenesulphonate salt, (S)-Tetrahydrofuran-3-amine-4-methylbenzenesulphonate, 3-Furanamine, tetrahydro-, (3S)-, 4-methylbenzenesulphonate (1:1)
OR322596-1G OR322596-5G
APOSOR322596-1GEA 1075 SEK
APOSOR322596-1G APOSOR322596-5G
(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulphonate salt 97%
(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulphonate salt
Formel: C₁₁H₁₇NO₄S
MW: 259.32 g/mol
Densitet: 1.012 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00066238
CAS nummer: 104530-79-2
UN: 1993
ADR: 3 III