4-Methylpyrimidine

Leverantör: Apollo Scientific

OR322543-10G OR322543-25G
APOSOR322543-10GEA 465 SEK
APOSOR322543-10G APOSOR322543-25G
4-Methylpyrimidine
4-Methylpyrimidine
Formel: C₅H₆N₂
MW: 94,12 g/mol
Kokpunkt: 141…142 °C
Densitet: 1,031 g/cm³ (25 °C)
Flampunkt: 40 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006115
CAS nummer: 3438-46-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR