4-(Methylthio)benzylamine
Leverantör: Apollo Scientific
Synonyms: (4-(Methylthio)phenyl)methanamine
OR322186-1G OR322186-5G
APOSOR322186-1GEA 1075 SEK
APOSOR322186-1G APOSOR322186-5G
4-(Methylthio)benzylamine
4-(Methylthio)benzylamine
Formel: C₈H₁₁NS
MW: 153,2500 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01310831
CAS nummer: 83171-39-5