Trityl alcohol

Leverantör: Apollo Scientific

OR322041-100G OR322041-500G
APOSOR322041-100GEA 290 SEK
APOSOR322041-100G APOSOR322041-500G
Trityl alcohol
Formel: C₁₉H₁₆O
Storage Temperature: Omgivande
MDL Number: MFCD00004445
CAS nummer: 76-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR