1-(Aminomethyl)cyclopentanol hydrochloride 97%
Leverantör: Apollo Scientific
OR321461-1G OR321461-5G
APOSOR321461-1GEA 1750 SEK
APOSOR321461-1G APOSOR321461-5G
1-(Aminomethyl)cyclopentanol hydrochloride 97%
1-(Aminomethyl)cyclopentanol hydrochloride
Formel: C₆H₁₄ClNO
MW: 151,64 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD10686694
CAS nummer: 76066-27-8