5-Bromoindan-2-ol 97%

Leverantör: Apollo Scientific
Varning

Synonyms: 5-Bromo-2,3-dihydro-1H-inden-2-ol

OR321364-1G OR321364-100MG OR321364-250MG
APOSOR321364-1GEA 25275 SEK
APOSOR321364-1G APOSOR321364-100MG APOSOR321364-250MG
5-Bromoindan-2-ol 97%
5-Bromoindan-2-ol
Formel: C₉H₉BrO
MW: 213,0700 g/mol
Smältpunkt: 115…117 °C
Storage Temperature: Omgivande
MDL Number: MFCD12755975
CAS nummer: 862135-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR