1-Boc-3-cyanopiperidine 97%

Leverantör: Apollo Scientific
Varning

Synonyms: tert-butyl 3-cyanopiperidine-1-carboxylate, 1-Boc-3-cyanopiperidine, 3-Cyano-1-piperidinecarboxylic acid tert-Butyl ester, tert-Butyl 3-Cyano-1-piperidinecarboxylate, 1-Boc-3-piperidinecarbonitrile, 1-tert-Butoxycarbonyl-3-piperidinecarbonitrile, 1-tert-Butoxycarbonyl-3-cyanopiperidine

OR321352-100MG OR321352-250MG OR321352-1G
APOSOR321352-100MGEA 8325 SEK
APOSOR321352-100MG APOSOR321352-250MG APOSOR321352-1G
1-Boc-3-cyanopiperidine 97%
1-Boc-3-cyanopiperidine
Formel: C₁₁H₁₈N₂O₂
MW: 210,27 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD01861222
CAS nummer: 91419-53-3
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR