4-Methoxy-3-(2-propynyloxy)benzenecarbaldehyde 95%
Leverantör: Apollo Scientific
OR32119-1G OR32119-5G
APOSOR32119-1GEA 2750 SEK
APOSOR32119-1G APOSOR32119-5G
4-Methoxy-3-(2-propynyloxy)benzenecarbaldehyde 95%
4-Methoxy-3-(2-propynyloxy)benzenecarbaldehyde
Formel: C₁₁H₁₀O₃
MW: 190,2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03001224
CAS nummer: 145654-01-9