2-(1,3,4-Oxadiazol-2-yl)phenol 95%
Leverantör: Apollo Scientific
OR32000-1G
OR32000-25G
OR32000-5G
APOSOR32000-1GEA
2075
SEK
APOSOR32000-1G
APOSOR32000-25G
APOSOR32000-5G
2-(1,3,4-Oxadiazol-2-yl)phenol 95%
2-(1,3,4-Oxadiazol-2-yl)phenol
Formel:
C₈H₆N₂O₂ MW: 162,15 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00020713 CAS nummer: 1008-65-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...