5-Bromo-7-nitroindoline 95+%

Leverantör: Apollo Scientific

OR318111-5G OR318111-25G
APOSOR318111-5GEA 1575 SEK
APOSOR318111-5G APOSOR318111-25G
5-Bromo-7-nitroindoline 95+%
5-Bromo-7-nitroindoline
Formel: C₈H₇BrN₂O₂
MW: 243.06 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00005708
CAS nummer: 80166-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR