4-Methyl-3-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)benzoic acid 95+%
Leverantör: Apollo Scientific
Varning
Synonyms: 4-Methyl-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)benzoic acid
OR318073-1G OR318073-10G OR318073-5G
APOSOR318073-1GEA 4100 SEK
APOSOR318073-1G APOSOR318073-10G APOSOR318073-5G
4-Methyl-3-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)benzoic acid 95+%
4-Methyl-3-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)benzoic acid
Formel: C₁₄H₁₉BO₄
MW: 262.1093 g/mol
Smältpunkt: 237…238 °C
Storage Temperature: Omgivande
MDL Number: MFCD09261433
CAS nummer: 515131-35-8