1-(Pyridin-2-yl)-1,4-diazepane hydrochloride 95+%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-(Pyridin-2-yl)homopiperazine hydrochloride
OR318071-1G OR318071-5G OR318071-10G
APOSOR318071-1GEA 3050 SEK
APOSOR318071-1G APOSOR318071-5G APOSOR318071-10G
1-(Pyridin-2-yl)-1,4-diazepane hydrochloride 95+%
1-(Pyridin-2-yl)-1,4-diazepane hydrochloride
Formel: C₁₀H₁₆ClN₃
MW: 213.71 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11840325
CAS nummer: 1177314-80-5