2-(tert-Butylcarbonyl)-1,2,3,4-tetrahydrobenzo[b][1,6]naphthyridine-10-carboxylic acid 95+%
Leverantör: Apollo Scientific
OR318053-1G OR318053-5G
APOSOR318053-1GEA 4125 SEK
APOSOR318053-1G APOSOR318053-5G
2-(tert-Butylcarbonyl)-1,2,3,4-tetrahydrobenzo[b][1,6]naphthyridine-10-carboxylic acid 95+%
2-(tert-Butylcarbonyl)-1,2,3,4-tetrahydrobenzo[b][1,6]naphthyridine-10-carboxylic acid
Formel: C₁₈H₂₀N₂O₃
Storage Temperature: Omgivande
MDL Number: MFCD03645391
CAS nummer: 887432-93-1