Ethyl-5-propyl-1H-pyrazole-3-carboxylate 95+%
Leverantör: Apollo Scientific
Synonyms: Ethyl 5-Propylpyrazole-3-carboxylate
OR318012-1G OR318012-25G OR318012-5G
APOSOR318012-1GEA 895 SEK
APOSOR318012-1G APOSOR318012-25G APOSOR318012-5G
Ethyl-5-propyl-1H-pyrazole-3-carboxylate 95+%
Ethyl-5-propyl-1H-pyrazole-3-carboxylate
Formel: C₉H₁₄N₂O₂
MW: 182,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08700607
CAS nummer: 92945-27-2