1-Oxa-6-azaspiro[3.3]heptane oxalate 98%
Leverantör: Apollo Scientific
OR317044-1G OR317044-100MG OR317044-250MG
APOSOR317044-1GEA 11900 SEK
APOSOR317044-1G APOSOR317044-100MG APOSOR317044-250MG
1-Oxa-6-azaspiro[3.3]heptane oxalate 98%
1-Oxa-6-azaspiro[3.3]heptane oxalate
Formel: C₇H₁₁NO₅
Storage Temperature: Omgivande
MDL Number: MFCD17214340
CAS nummer: 936947-34-1