2-Bromodibenzothiophene 99%

Leverantör: Apollo Scientific
Varning

Synonyms: 2-Bromodibenzo[b,d]thiophene

OR31664-25G OR31664-5G
APOSOR31664-25GEA 415 SEK
APOSOR31664-25G APOSOR31664-5G
2-Bromodibenzothiophene 99%
2-Bromodibenzothiophene
Formel: C₁₂H₇BrS
MW: 263,15 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00089285
CAS nummer: 22439-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR