D-allo-Threonine
Leverantör: Apollo Scientific
Varning
Synonyms: H-D-Allo-Thr-OH, (2R,3R)-2-Amino-3-hydroxybutanoic acid
OR316027-1G OR316027-25G OR316027-5G
APOSOR316027-1GEA 685 SEK
APOSOR316027-1G APOSOR316027-25G APOSOR316027-5G
D-allo-Threonine
D-allo-Threonine
Formel: C₄H₉NO₃
MW: 119,12 g/mol
Smältpunkt: 268…276 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00004526
CAS nummer: 24830-94-2
EINECS: 246-488-0