N-Benzyloxycarbonyl-L-alaninol

Leverantör: Apollo Scientific

Synonyms: Z-Ala-ol, (S)-Benzyl (1-hydroxypropan-2-yl)carbamate, (S)-(2-Hydroxy-1-methylethyl)carbamic acid benzyl ester, N-Cbz-L-Alaninol

OR316022-25G OR316022-5G
APOSOR316022-25GEA 1300 SEK
APOSOR316022-25G APOSOR316022-5G
N-Benzyloxycarbonyl-L-alaninol
N-Cbz-L-Alaninol
Formel: C₁₁H₁₅NO₃
MW: 209,25 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00672530
CAS nummer: 66674-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR