3-Methoxypropionic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 3-Methoxypropanoic acid
OR315601-100G OR315601-10G OR315601-25G
APOSOR315601-100GEA 6425 SEK
APOSOR315601-100G APOSOR315601-10G APOSOR315601-25G
3-Methoxypropionic acid
3-Methoxypropionic acid
Formel: C₄H₈O₃
MW: 104,1100 g/mol
Kokpunkt: 116 °C
Densitet: 1,108 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00042654
CAS nummer: 2544-06-1