Ethyl-3-oxocyclobutanecarboxylate

Leverantör: Apollo Scientific
Varning

Synonyms: Ethyl 3-oxocyclobutanecarboxylate

OR315578-1G OR315578-25G OR315578-5G
APOSOR315578-1GEA 620 SEK
APOSOR315578-1G APOSOR315578-25G APOSOR315578-5G
Ethyl-3-oxocyclobutanecarboxylate
Ethyl-3-oxocyclobutanecarboxylate
Formel: C₇H₁₀O₃
MW: 142,154 g/mol
Kokpunkt: 86…90 °C/0,6mmHg
Storage Temperature: Kylskåp
MDL Number: MFCD09834114
CAS nummer: 87121-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR