2-Bromotriphenylene 97%

Leverantör: Apollo Scientific
Varning

OR315396-25G OR315396-5G OR315396-1G
APOSOR315396-25GEA 2550 SEK
APOSOR315396-25G APOSOR315396-5G APOSOR315396-1G
2-Bromotriphenylene 97%
2-Bromotriphenylene
Formel: C₁₈H₁₁Br
Storage Temperature: Omgivande
MDL Number: MFCD16659043
CAS nummer: 19111-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR