(S)-(+)-2-Methylbutyric acid 98%
Leverantör: Apollo Scientific
Fara
Synonyms: (S)-(+)-2-Methylbutanoic acid, (S)-2-Methylbutanoic acid
OR315294-1G OR315294-5G OR315294-250MG
APOSOR315294-1GEA 1475 SEK
APOSOR315294-1G APOSOR315294-5G APOSOR315294-250MG
(S)-(+)-2-Methylbutyric acid 98%
(S)-(+)-2-Methylbutyric acid
Formel: C₅H₁₀O₂
MW: 102,13 g/mol
Kokpunkt: 78 °C/15mmHg
Densitet: 0,938 g/cm³ (25 °C)
Flampunkt: 73 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00063166
CAS nummer: 1730-91-2
UN: 3265
ADR: 8,II