1-Bromo-3-chloro-5-nitrobenzene
Leverantör: Apollo Scientific
Varning
OR315204-1G OR315204-5G
APOSOR315204-1GEA 260 SEK
APOSOR315204-1G APOSOR315204-5G
1-Bromo-3-chloro-5-nitrobenzene
1-Bromo-3-chloro-5-nitrobenzene
Formel: C₆H₃BrClNO₂
MW: 236,4500 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08275708
CAS nummer: 219817-43-3