5-Ethyl-2-methylpyridine
Leverantör: Apollo Scientific
|
Fara
|
OR3152-25G
OR3152-100G
APOSOR3152-25GEA
210
SEK
APOSOR3152-25G
APOSOR3152-100G
5-Ethyl-2-methylpyridine
5-Ethyl-2-methylpyridine
Formel:
C₈H₁₁N MW: 121,18 g/mol Kokpunkt: 178 °C Smältpunkt: –71 °C Densitet: 0,919 g/cm³ (25 °C) Flampunkt: 70 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00006344 CAS nummer: 104-90-5 EINECS: 203-250-0 UN: 2300 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...