6-Methoxy-2-naphthylacetic acid 97+%
Leverantör: Apollo Scientific
Synonyms: 2-(6-Methoxynaphthalen-2-yl)acetic acid
OR315155-1G OR315155-250MG OR315155-5G
APOSOR315155-1GEA 1425 SEK
APOSOR315155-1G APOSOR315155-250MG APOSOR315155-5G
6-Methoxy-2-naphthylacetic acid 97+%
6-Methoxy-2-naphthylacetic acid
Formel: C₁₃H₁₂O₃
MW: 216.236 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00033105
CAS nummer: 23981-47-7