4,5,6,7-Tetrahydro-1H-benzo[d]imidazole-5-carboxylic acid hydrochloride 95%
Leverantör: Apollo Scientific
OR315144-25G OR315144-5G
APOSOR315144-25GEA 5600 SEK
APOSOR315144-25G APOSOR315144-5G
4,5,6,7-Tetrahydro-1H-benzo[d]imidazole-5-carboxylic acid hydrochloride 95%
4,5,6,7-Tetrahydro-1H-benzo[d]imidazole-5-carboxylic acid hydrochloride
Formel: C₈H₁₁ClN₂O₂
MW: 202.6381 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18157694
CAS nummer: 131020-57-0