2-Chloro-4-chloromethylpyridine 97%

Leverantör: Apollo Scientific
Varning

Synonyms: 2-Chloro-4-(chloromethyl)pyridine

OR315110-25G OR315110-5G
APOSOR315110-25GEA 4875 SEK
APOSOR315110-25G APOSOR315110-5G
2-Chloro-4-chloromethylpyridine 97%
2-Chloro-4-chloromethylpyridine
Formel: C₆H₅Cl₂N
MW: 162,02 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09991655
CAS nummer: 101990-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR