5-Amino-3-bromo-2-methylpyridine

Leverantör: Apollo Scientific
Varning

OR315100-1G OR315100-5G
APOSOR315100-1GEA 290 SEK
APOSOR315100-1G APOSOR315100-5G
5-Amino-3-bromo-2-methylpyridine
5-Amino-3-bromo-2-methylpyridine
Formel: C₆H₇BrN₂
Storage Temperature: Kylskåp
MDL Number: MFCD08060462
CAS nummer: 186593-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR