4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride
Leverantör: Apollo Scientific
OR315069-100G OR315069-25G OR315069-5G
APOSOR315069-100GEA 805 SEK
APOSOR315069-100G APOSOR315069-25G APOSOR315069-5G
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride
Formel: C₇H₁₀ClNS
MW: 175,68 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05861485
CAS nummer: 28783-41-7