Ferroceneboronic acid 95%
Leverantör: Apollo Scientific
Varning
OR315060-1G OR315060-5G
APOSOR315060-1GEA 1100 SEK
APOSOR315060-1G APOSOR315060-5G
Ferroceneboronic acid 95%
Ferroceneboronic acid
Formel: C₁₀H₁₁BFeO₂
MW: 229,85 g/mol
Smältpunkt: 145…150 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00059074
CAS nummer: 12152-94-2
EINECS: 235-272-1